missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylamine, 95%

Product Code. 11491507
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491507 5 g 5g
11401517 25 g 25g
11411517 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491507 Supplier Thermo Scientific Alfa Aesar Supplier No. B24963.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10269-01-9 | C7H8BrN | 186.052 g/mol

3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10269-01-9
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD01026119
InChI Key SUYJXERPRICYRX-UHFFFAOYSA-N
Synonym 3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109
PubChem CID 457587
IUPAC Name (3-bromophenyl)methanamine
SMILES C1=CC(=CC(=C1)Br)CN

Specifications

Odor Amine-like
Refractive Index 1.5885
Quantity 5 g
UN Number UN2735
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 186.06
Percent Purity 97%
Chemical Name or Material 3-Bromobenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.