missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylamine, 95%

Product Code. 10159679 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10159679 25 g Glass bottle 25g
10078269 1 g Glass bottle 1g
10627970 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10159679 Supplier Thermo Scientific Acros Supplier No. 428960250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10269-01-9 | C7H8BrN | 186.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10269-01-9
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD01026119
InChI Key SUYJXERPRICYRX-UHFFFAOYSA-N
Synonym 3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109
PubChem CID 457587
IUPAC Name (3-bromophenyl)methanamine
SMILES C1=CC(=CC(=C1)Br)CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2NH2
Refractive Index 1.5850 to 1.5900 (20°C, 589nm)
Quantity 25 g
Formula Weight 186.05
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Bromobenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.