missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzyl bromide, 99%

Product Code. 11497367
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497367 10 g 10g
11407377 50 g 50g
11417377 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497367 Supplier Thermo Scientific Alfa Aesar Supplier No. A15075.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 823-78-9 | C7H6Br2 | 249.933 g/mol

3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-78-9
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000176
InChI Key ZPCJPJQUVRIILS-UHFFFAOYSA-N
Synonym 3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
PubChem CID 69979
IUPAC Name 1-bromo-3-(bromomethyl)benzene
SMILES C1=CC(=CC(=C1)Br)CBr

Specifications

Melting Point 40°C to 44°C
Density 1.56
Boiling Point 128°C to 130°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN1759
Beilstein 2078683
Solubility Information Insoluble in water.
Formula Weight 249.94
Percent Purity 99%
Chemical Name or Material 3-Bromobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3-Bromobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.