missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoyl chloride, 98%

Product Code. 11402308
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11402308

Brand: Thermo Scientific Alfa Aesar A15256.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1711-09-7 | C7H4BrClO | 219.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-09-7
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00000669
InChI Key PBOOZQFGWNZNQE-UHFFFAOYSA-N
Synonym m-bromobenzoyl chloride, benzoyl chloride, 3-bromo, 3-bromobenzoylchloride, benzoyl chloride, m-bromo, 3-bromobenzoic acid chloride, attercop-chm at106527, 3-brom-benzoylchlorid, 3-bromo-benzoylchlorid, 3-bromobenzoic chloride, pubchem13574
PubChem CID 74377
IUPAC Name 3-bromobenzoyl chloride
SMILES C1=CC(=CC(=C1)Br)C(=O)Cl

Specifications

Density 1.666
Boiling Point 247°C to 251°C
Flash Point 107°C (224°F)
Refractive Index 1.595
Quantity 10 g
UN Number UN3265
Beilstein 1635505
Sensitivity Moisture sensitive
Formula Weight 219.47
Percent Purity 98%
Chemical Name or Material 3-Bromobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : 3-Bromobenzoyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.