missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzonitrile, 99%

Product Code. 10675902 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10675902 10 g Glass bottle 10g
10658074 2.5 g Glass bottle 2.5g
10414955 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10675902 Supplier Thermo Scientific Acros Supplier No. 106700100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6952-59-6 | C7H4BrN | 182.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6952-59-6
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD00001796
InChI Key STXAVEHFKAXGOX-UHFFFAOYSA-N
Synonym m-bromobenzonitrile, benzonitrile, 3-bromo, benzonitrile, m-bromo, 3-cyanobromobenzene, 1-bromo-3-cyanobenzene, 3-bromobenzenecarbonitrile, 3-bromo benzonitrile, 3-bromobenzonitride, m-brombenzonitril, 3-brombenzonitrile
PubChem CID 23381
IUPAC Name 3-bromobenzonitrile
SMILES BrC1=CC=CC(=C1)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 37°C to 41°C
Color Beige or White to Yellow
Boiling Point 225°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CN
Quantity 10 g
Beilstein 09, 350
Formula Weight 182.02
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material 3-Bromobenzonitrile, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.