missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoic Acid, TRC

Product Code. 30330848
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330848 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330848 Supplier Toronto Research Chemicals Supplier No. TRCB68050550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromobenzoic Acid
CAS 585-76-2
Molecular Formula C7H5BrO2
Quantity 50 g
Synonym m-Bromobenzoic Acid, NSC 176129, NSC 32739, NSC 3980, NSC 83519
InChI Formula InChI=1S/C7H5BrO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,(H,9,10)
SMILES OC(=O)c1cccc(Br)c1
IUPAC Name 3-bromobenzoic acid
Molecular Weight (g/mol) 201.02
Formula Weight 199.9473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.