missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoic acid, 99%

Product Code. 10590344 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10590344 100 g Plastic bottle 100g
10289313 5 g Glass Bottle 5g
10032300 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10590344 Supplier Thermo Scientific Acros Supplier No. 181351000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 585-76-2 | C7H5BrO2 | 201.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-76-2
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00002487
InChI Key VOIZNVUXCQLQHS-UHFFFAOYSA-N
Synonym m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid
PubChem CID 11456
IUPAC Name 3-bromobenzoic acid
SMILES C1=CC(=CC(=C1)Br)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 154°C to 158°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula BrC6H4CO2H
Quantity 100 g
Beilstein 09, 349
Solubility Information Solubility in water: 0.54g/L (32°C)
Formula Weight 201.02
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Bromobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.