missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzoic acid, 98+%

Product Code. 11414344
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414344 100 g 100g
11404344 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414344 Supplier Thermo Scientific Alfa Aesar Supplier No. A12606.22
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 585-76-2 | C7H5BrO2 | 201.019 g/mol

3-Bromobenzoic acid was used as test solute in the determination of acidity constants by capillary zone electrophoresis. It was used as internal standard to study the retention mechanisms of an unmodified and a hydroxylated polystyrene-divinylbenzene polymer by solid-phase extraction. It was also used in synthesis of N-(1,1-dimethyl-2-hydroxyethyl)-3-bromobenzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzoic acid was used as test solute in the determination of acidity constants by capillary zone electrophoresis. It was used as internal standard to study the retention mechanisms of an unmodified and a hydroxylated polystyrene-divinylbenzene polymer by solid-phase extraction. It was also used in synthesis of N-(1,1-dimethyl-2-hydroxyethyl)-3-bromobenzamide.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-76-2
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00002487
InChI Key VOIZNVUXCQLQHS-UHFFFAOYSA-N
Synonym m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid
PubChem CID 11456
IUPAC Name 3-bromobenzoic acid
SMILES C1=CC(=CC(=C1)Br)C(=O)O

Specifications

Melting Point 153°C to 157°C
Density 1.948
Quantity 100 g
Beilstein 2041792
Solubility Information Insoluble in water.
Formula Weight 201.02
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.