missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzeneboronic acid N-methyldiethanolamine ester, 98%, Thermo Scientific™

Catalog Number 11394765
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
374538-00-8
C11H15BBrNO2
283.96
MFCD02093962
CYGPBFLFZQMUBN-UHFFFAOYSA-N
2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaborocane, 3-bromobenzeneboronic acid n-methyldiethanolamine ester, 2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaboraperhydroocine, 3-bromophenylboronic acid n-methyl diethanolamine cyclic este
10039503
2-(3-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
B1(OCCN(CCO1)C)C2=CC(=CC=C2)Br
This item is not returnable. View return policy
374538-00-8
C11H15BBrNO2
283.96
MFCD02093962
CYGPBFLFZQMUBN-UHFFFAOYSA-N
2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaborocane, 3-bromobenzeneboronic acid n-methyldiethanolamine ester, 2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaboraperhydroocine, 3-bromophenylboronic acid n-methyl diethanolamine cyclic este
10039503
2-(3-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
B1(OCCN(CCO1)C)C2=CC(=CC=C2)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 128°C to 129°C
Quantity 1 g
Formula Weight 283.96
Percent Purity 98%
Chemical Name or Material 3-Bromobenzeneboronic acid N-methyldiethanolamine ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'