missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzene-1-sulfonamide, 97%, Thermo Scientific™

Product Code. 10550553
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 89599-01-9
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.083
MDL Number MFCD00084903
InChI Key MUBJNMWVQGHHLG-UHFFFAOYSA-N
Synonym 3-bromobenzenesulphonamide, 3-bromobenzene-1-sulfonamide, benzenesulfonamide, 3-bromo, 3-bromo-benzenesulfonamide, 3-bromobenesulfonamide, 3-bromo-benzenesulfonic acid amide, 3-bromanylbenzenesulfonamide, pubchem11759, 3-bromophenylsulfonamide, m-bromobenzenesulfonamide
PubChem CID 2734765
IUPAC Name 3-bromobenzenesulfonamide
SMILES C1=CC(=CC(=C1)Br)S(=O)(=O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula BrC6H4SO2NH2
Quantity 10 g
Formula Weight 236.09
Percent Purity 97%
Chemical Name or Material 3-Bromobenzene-1-sulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'