missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzene-1-sulfonamide, 97%, Thermo Scientific™

Product Code. 10275464
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10275464 25 g 25g
10293282 1 g 1g
10550553 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10275464 Supplier Thermo Scientific Maybridge Supplier No. TL00520EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89599-01-9
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.083
MDL Number MFCD00084903
InChI Key MUBJNMWVQGHHLG-UHFFFAOYSA-N
Synonym 3-bromobenzenesulphonamide, 3-bromobenzene-1-sulfonamide, benzenesulfonamide, 3-bromo, 3-bromo-benzenesulfonamide, 3-bromobenesulfonamide, 3-bromo-benzenesulfonic acid amide, 3-bromanylbenzenesulfonamide, pubchem11759, 3-bromophenylsulfonamide, m-bromobenzenesulfonamide
PubChem CID 2734765
IUPAC Name 3-bromobenzenesulfonamide
SMILES C1=CC(=CC(=C1)Br)S(=O)(=O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula BrC6H4SO2NH2
Quantity 25 g
Formula Weight 236.09
Percent Purity 97%
Chemical Name or Material 3-Bromobenzene-1-sulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.