missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzanthrone, tech. 85%

Product Code. 11391907
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391907 100 g 100g
11381907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391907 Supplier Thermo Scientific Alfa Aesar Supplier No. L07422.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81-96-9 | C17H9BrO | 309.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-96-9
Molecular Formula C17H9BrO
Molecular Weight (g/mol) 309.162
MDL Number MFCD00021094
InChI Key WVECFEIAZAKUNF-UHFFFAOYSA-N
Synonym 3-bromobenzanthrone, 3-brombenzanthrone, benzanthrone, 3-bromo, 7-bromomesobenzanthrone, 3-bromo-7h-benzo de anthracen-7-one, 7h-benz de anthracen-7-one, 3-bromo, 3-bromobenz d,e anthrone, 3-bromo-7h-benz de anthracen-7-one, 3-bromo-7h-benz de anthracene-7-one, unii-3dh9916fno
PubChem CID 6696
IUPAC Name 3-bromobenzo[b]phenalen-7-one
SMILES C1=CC=C2C(=C1)C3=C4C(=C(C=C3)Br)C=CC=C4C2=O

Specifications

Melting Point 164°C to 168°C
Quantity 100 g
Beilstein 1975512
Formula Weight 309.17
Percent Purity 85%
Grade Technical
Chemical Name or Material 3-Bromobenzanthrone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.