missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzamide, 98+%

Product Code. 15401979
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401979 25 g 25g
15411979 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401979 Supplier Thermo Scientific Alfa Aesar Supplier No. H64672.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22726-00-7 | C7H6BrNO | 200.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22726-00-7
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00017127
InChI Key ODJFDWIECLJWSR-UHFFFAOYSA-N
Synonym m-bromobenzamide, benzamide, 3-bromo, 3-bromo-benzamide, benzamide, m-bromo, 3-carbamoyl bromobenzene, 3-bromanylbenzamide, pubchem3734, 3-bromophenylcarboxamide, acmc-209fyc, 3-bromobenzoic acid amide
PubChem CID 89807
IUPAC Name 3-bromobenzamide
SMILES C1=CC(=CC(=C1)Br)C(=O)N

Specifications

Melting Point 156°C to 159°C
Color White
Quantity 25 g
Beilstein 1860187
Formula Weight 200.04
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 3-Bromobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.