missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzaldehyde, 97%

Product Code. 11463934
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463934 500 g 500g
11443934 25 g 25g
11453934 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463934 Supplier Thermo Scientific Alfa Aesar Supplier No. A11941.36
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3132-99-8 | C7H5BrO | 185.02 g/mol

A substituted benzaldehyde used in the synthesis of more complex pharmaceutical compounds. It is also employed as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A substituted benzaldehyde used in the synthesis of more complex pharmaceutical compounds. It is also employed as an active pharmaceutical ingredient.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Keep away from oxidizing agents and air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3132-99-8
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003345
InChI Key SUISZCALMBHJQX-UHFFFAOYSA-N
Synonym m-bromobenzaldehyde, benzaldehyde, 3-bromo, benzaldehyde, m-bromo, 3-bromobenzeldehyde, unii-w0ard2zj82, 3-bromo benzaldehyde, 3-bromo-benzaldehyde, w0ard2zj82, benzaldehyde, m-bromo-6ci,7ci,8ci, 3-bromobenzaidehyde
PubChem CID 76583
IUPAC Name 3-bromobenzaldehyde
SMILES C1=CC(=CC(=C1)Br)C=O

Specifications

Melting Point 18°C to 21°C
Density 1.58
Boiling Point 234°C to 236°C
Flash Point 96°C (204°F)
Odor Characteristic
Refractive Index 1.594
Quantity 500 g
Beilstein 1634534
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 185.03
Percent Purity 97%
Chemical Name or Material 3-Bromobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.