missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzaldehyde, 96%

Product Code. 10645522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10645522 25 mL Glass bottle 25mL
10656672 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10645522 Supplier Thermo Scientific Acros Supplier No. 106660250
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 3132-99-8 | C7H5BrO | 185.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3132-99-8
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003345
InChI Key SUISZCALMBHJQX-UHFFFAOYSA-N
Synonym m-bromobenzaldehyde, benzaldehyde, 3-bromo, benzaldehyde, m-bromo, 3-bromobenzeldehyde, unii-w0ard2zj82, 3-bromo benzaldehyde, 3-bromo-benzaldehyde, w0ard2zj82, benzaldehyde, m-bromo-6ci,7ci,8ci, 3-bromobenzaidehyde
PubChem CID 76583
IUPAC Name 3-bromobenzaldehyde
SMILES C1=CC(=CC(=C1)Br)C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 18.0°C to 21.0°C
Color Colorless to Yellow
Density 1.5800g/mL
Boiling Point 228.0°C to 230.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CHO
Refractive Index 1.5920 to 1.5960
Quantity 25 mL
Beilstein 07, 238
Specific Gravity 1.58
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in most organic solvents
Formula Weight 185.02
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material 3-Bromobenzaldehyde, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.