missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromoanisole, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 3-Bromoanisole |
| CAS | 2398-37-0 |
| Molecular Formula | C7 H7 Br O |
| Quantity | 25 g |
| Synonym | 3-Bromoanisole, Benzene, 1-bromo-3-methoxy- (9CI, ACI), 1-Bromo-3-methoxybenzene (ACI), Anisole, m-bromo- (6CI, 7CI, 8CI), 3-Bromoanisole, 3-Bromobenzene methyl ether, 3-Methoxy-1-bromobenzene, 3-Methoxybromobenzene, 3-Methoxyphenyl bromide, m-Anisyl bromide, m-Bromoanisole, m-Bromoanisole, m-Bromomethoxybenzene, m-Bromophenyl methyl ether, m-Methoxybromobenzene, m-Methoxyphenyl bromide, NSC 82293 |
| InChI Formula | InChI=1S/C7H7BrO/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3 |
| SMILES | COc1cccc(Br)c1 |
| IUPAC Name | 1-bromo-3-methoxybenzene |
| Molecular Weight (g/mol) | 187.03 |
| Formula Weight | 185.968 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction