missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoadamantane-1-carboxylic acid, 97%

Product Code. 15477058
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477058 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15477058 Supplier Thermo Scientific Alfa Aesar Supplier No. H60580.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 21816-08-0 | C11H15BrO2 | 259.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21816-08-0
Melting Point 146°C to 150°C
Molecular Formula C11H15BrO2
MDL Number MFCD00167820
Quantity 1 g
Synonym 1-bromo-3-adamantanecarboxylic acid, 3-bromo-1-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, 3-bromo, 3-bromo-adamantane-1-carboxylic acid, 3-adamantanecarboxylic acid, 1-bromo, 3-bromoadamantanecarboxylic acid, 3-bromotricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, acmc-1cnbw, maybridge1_005590, 5s,7r-3-bromoadamantane-1-carboxylate
InChI Key DJUDQBVINJIMFO-UHFFFAOYSA-N
SMILES C1C2CC3(CC1CC(C2)(C3)Br)C(=O)O
IUPAC Name 3-bromoadamantane-1-carboxylic acid
Molecular Weight (g/mol) 259.143
PubChem CID 30818
Formula Weight 259.14
Percent Purity 97%
Chemical Name or Material 3-Bromoadamantane-1-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.