missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromoacetophenone, 97%

Product Code. 10695132 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10695132 25 g Glass bottle 25g
10323522 5 g Glass bottle 5g
10474765 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10695132 Supplier Thermo Scientific Acros Supplier No. 106580250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2142-63-4 | C8H7BrO | 199.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2142-63-4
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000083
InChI Key JYAQYXOVOHJRCS-UHFFFAOYSA-N
Synonym 3'-bromoacetophenone, m-bromoacetophenone, 1-3-bromophenyl ethanone, 1-3-bromophenyl ethan-1-one, ethanone, 1-3-bromophenyl, 1-acetyl-3-bromobenzene, 3-bromoacetophenone, 1-3-bromo-phenyl-ethanone, 3-bromo acetophenone, acetophenone, 3'-bromo
PubChem CID 16502
IUPAC Name 1-(3-bromophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.4900g/mL
Boiling Point 79.0°C to 81.0°C (2.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula BrC6H4COCH3
Refractive Index 1.5749 to 1.5769
Quantity 25 g
Specific Gravity 1.49
Formula Weight 199.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3'-Bromoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 3'-Bromoacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.