missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-N-methylaniline, 97%, Thermo Scientific Chemicals

Product Code. 15441718
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441718 25 g 25g
15421718 1 g 1g
15431718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441718 Supplier Thermo Scientific Alfa Aesar Supplier No. H55464.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66584-32-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD05664376
InChI Key HKOSFZXROYRVJT-UHFFFAOYSA-N
Synonym n-methyl-3-bromoaniline, 3-bromo-n-methyl aniline, benzenamine, 3-bromo-n-methyl, acmc-20an89, 3-bromophenyl-methylamine, 3-bromo-phenyl-methyl-amine, 3-bromo-n-methylaniline
PubChem CID 7018299
IUPAC Name 3-bromo-N-methylaniline
SMILES CNC1=CC(=CC=C1)Br

Specifications

Color Colorless to Yellow
Density 1.461 g/mL
Boiling Point 253°C
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.621
Quantity 25 g
Formula Weight 186.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-N-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.