missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Bromo-N-(2-iodophenyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H60408.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
3-Bromo-N-(2-iodophenyl)benzenesulfonamide | |
C12H9BrINO2S | |
438.08 | |
MFCD06962762 |
Specifications
3-Bromo-N-(2-iodophenyl)benzenesulfonamide | |
C12H9BrINO2S | |
MFCD06962762 |
97% | |
438.08 | |
1g |