missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride, 97%

Product Code. 11395545
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395545 5 g 5g
11385545 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395545 Supplier Thermo Scientific Alfa Aesar Supplier No. L19711.06
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 351003-46-8 | C7H3BrClF3O2S | 323.508 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-46-8
Molecular Formula C7H3BrClF3O2S
Molecular Weight (g/mol) 323.508
MDL Number MFCD03094393
InChI Key YBUJCZFWJSUTSN-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl benzenesulfonyl chloride, 3-bromo-5-trifluoromethyl benzene-1-sulfonyl chloride, 3-bromo-5-trifluoromethyl benzenesulphonylchloride, 3-bromo-5-trifluoromethyl benzene sulphonyl chloride, 5-bromo-3-trifluoromethyl phenyl chlorosulfone, 3-bromo-5-trifluoromethylphenylsulphonyl chloride, 3-bromo-5-trifluoromethyl benzenesulphonyl chloride, benzenesulfonylchloride, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethylphenylsulfonyl chloride, 3-bromo-5-trifluoromethyl benzenesulfonylchloride
PubChem CID 2778610
IUPAC Name 3-bromo-5-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=C(C=C(C=C1S(=O)(=O)Cl)Br)C(F)(F)F

Specifications

Density 1.835
Boiling Point 217°C to 218°C
Flash Point 66°C (150°F)
Refractive Index 1.524
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 323.52
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.