missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)aniline (3-Amino-5-bromobenzotrifluoride), Mikromol™

Product Code. 30050500
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050500 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050500 Supplier Mikromol Supplier No. MM3374.090100
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Bromo-5-(trifluoromethyl)aniline (3-Amino-5-bromobenzotrifluoride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromo-5-(trifluoromethyl)aniline
CAS 54962-75-3
Analyte or Component Names 3-Bromo-5-(trifluoromethyl)aniline
Shipping Condition Room Temperature
Molecular Formula C7 H5 Br F3 N
Certifications/Compliance ISO 9001
Synonym Benzenamine, 3-bromo-5-(trifluoromethyl)-, 3-Bromo-5-(trifluoromethyl)benzenamine, 3-Amino-5-bromo-1-trifluoromethylbenzene, 3-Amino-5-bromobenzotrifluoride, 3-Bromo-5-(trifluoromethyl)aniline, 3-Bromo-5-(trifluoromethyl)phenylamine
InChI Formula InChI=1S/C7H5BrF3N/c8-5-1-4(7(9,10)11)2-6(12)3-5/h1-3H,12H2
SMILES Nc1cc(Br)cc(c1)C(F)(F)F
IUPAC Name 3-bromo-5-(trifluoromethyl)aniline
Molecular Weight (g/mol) 240.0205
Quantity 100 mg
Formula Weight 238.9557 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.