missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-nitrobenzaldehyde, ≥97%, Thermo Scientific Chemicals

Product Code. 15402149
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355134-13-3
Molecular Formula C7H4BrNO3
Molecular Weight (g/mol) 230.017
MDL Number MFCD00100097
InChI Key OUDCOMBHRXKPIJ-UHFFFAOYSA-N
Synonym benzaldehyde, 3-bromo-5-nitro, 3-bromo-5-nitro-benzaldehyde, 3-bromo-5-nitro benzaldehyde, ksc497m5f, 3-nitro-5-bromo-benzaldehyde, 3-bromanyl-5-nitro-benzaldehyde, 5-bromo-3-nitrobenzaldehyde, 3-bromo-5-nitrobenzaldehyde
PubChem CID 16743966
IUPAC Name 3-bromo-5-nitrobenzaldehyde
SMILES C1=C(C=C(C=C1[N+](=O)[O-])Br)C=O

Specifications

Melting Point 101°C to 106°C
Color Brown
Quantity 250 mg
Beilstein 3253229
Formula Weight 230.02
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 3-Bromo-5-nitrobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'