missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methylaniline, 98%

Product Code. 15478078
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478078 1 g 1g
15488078 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478078 Supplier Thermo Scientific Alfa Aesar Supplier No. H61519.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 74586-53-1 | C7H8BrN | 186.052 g/mol

3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74586-53-1
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD11845553
InChI Key YIZRPAWCIFTHNA-UHFFFAOYSA-N
Synonym 3-bromo-5-methylbenzenamine, 3-amino-5-bromotoluene, benzenamine, 3-bromo-5-methyl, 5-brom-3-toluidin, 5-bromo-m-toluidine, acmc-209ova, 3-bromo-5-methyl-aniline, 3-bromo-5-methyl aniline, ksc494e0l
PubChem CID 3018526
IUPAC Name 3-bromo-5-methylaniline
SMILES CC1=CC(=CC(=C1)Br)N

Specifications

Color Brown
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 186.05
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Bromo-5-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.