missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-iodopyridine, 95%

Product Code. 15481019
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481019 5 g 5g
15491019 250 mg 250mg
15471019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481019 Supplier Thermo Scientific Alfa Aesar Supplier No. H64007.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 233770-01-9 | C5H3BrIN | 283.894 g/mol

Reactant in the synthesis chiral 4, 4?-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis chiral 4, 4′-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 233770-01-9
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.894
MDL Number MFCD03086019
InChI Key AOOZLVWDZUPEHT-UHFFFAOYSA-N
PubChem CID 820409
IUPAC Name 3-bromo-5-iodopyridine
SMILES C1=C(C=NC=C1I)Br

Specifications

Melting Point 127°C to 131°C
Color White
Quantity 5 g
Beilstein 108478
Sensitivity Light sensitive
Synonym 3-bromo-5-iodo-pyridine, 5-bromo-3-iodopyridine, pyridine, 3-bromo-5-iodo, 3-bromo-5-iodpyridine, 3-iodo-5-bromopyridine, pubchem8080, acmc-1cdm9, pyridine,3-bromo-5-iodo, 3-bromo-5-iodopyridine, abbypharma ap-13-5787
Solubility Information Slightly soluble in water.
Formula Weight 283.89
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Bromo-5-iodopyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.