missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluoropyridine, 97%

Product Code. 10371330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10371330 1 g 1g
10199779 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10371330 Supplier Thermo Scientific Acros Supplier No. 437420010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 407-20-5 | C5H3BrFN | 175.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-20-5
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.99
MDL Number MFCD04112555
InChI Key HNNNBQRRIHKFLI-UHFFFAOYSA-N
Synonym 5-bromo-3-fluoropyridine, 3-bromo-5-fluoro-pyridine, 3-brom-5-fluorpyridin, 3-bromo-5-fluoro pyridine, pyridine, 3-bromo-5-fluoro, 3-bromo-5-fluorpyridine, 5-bromo-3-fluoride pyridine, 3-bromine-5-fluoride pyridine, pubchem3964, 3-bromo-5-fluoropyridin
PubChem CID 820206
IUPAC Name 3-bromo-5-fluoropyridine
SMILES FC1=CN=CC(Br)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 24.0°C to 28.0°C
Color Colorless to White
Boiling Point 78.0°C (11.0 Torr)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 175.99
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3-Bromo-5-fluoropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromo-5-fluoropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.