missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzyl bromide, 95%, Thermo Scientific™

Product Code. p-7058175
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410639 1 g 1g
15420639 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410639 Supplier Thermo Scientific Alfa Aesar Supplier No. H63769.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Bromo-5-fluorobenzyl bromide is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water (2.9 g/L) (25°C).

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216755-57-6
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.92
MDL Number MFCD09258959
InChI Key DAUWIPUGOIFZNF-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorobenzyl bromide, 1-bromo-3-bromomethyl-5-fluorobenzene, 3-fluoro-5-bromobenzyl bromide, 5-bromo-3-fluorobenzyl bromide, 3-fluoro-5-bromobenzylbromide, 5-bromo-3-bromomethyl-1-fluorobenzene, pubchem4886, acmc-20e3gr, 3-bromo-5-fluorobenzylbromide, benzene,1-bromo-3-bromomethyl-5-fluoro
PubChem CID 23509619
IUPAC Name 1-bromo-3-(bromomethyl)-5-fluorobenzene
SMILES FC1=CC(Br)=CC(CBr)=C1

Specifications

Quantity 1 g
UN Number UN3261
Solubility Information Slightly soluble in water (2.9g/L) (25°C).
Formula Weight 267.92
Percent Purity 95%
Chemical Name or Material 3-Bromo-5-fluorobenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.