missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Bromo-5-fluorobenzaldehyde, 95%

Product Code. 15451069
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15451069

Brand: Thermo Scientific Chemicals H64042.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Form: solid | CAS: 188813-02-7 | C7H4BrFO | 203.01 g/mol

3-Bromo-5-fluorobenzaldehyde is an intermediate in organic syntheses. Reatant in the synthesis of 3,4-diamino-3-cyclobutene-1,2-diones, 9,10-Dihydro-9,10-diboraanthracenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-fluorobenzaldehyde is an intermediate in organic syntheses. Reatant in the synthesis of 3,4-diamino-3-cyclobutene-1,2-diones, 9,10-Dihydro-9,10-diboraanthracenes.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air , heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188813-02-7
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD04116319
InChI Key MEFQRXHVMJPOKZ-UHFFFAOYSA-N
Synonym 3-fluoro-5-bromobenzaldehyde, benzaldehyde, 3-bromo-5-fluoro, 5-bromo-3-fluorobenzaldehyde, 3-bromo-5-fluoro-benzaldehyde, pubchem1420, acmc-209ers, ksc174q7p, 3-bromo-5-fluorobenzaldehyde
PubChem CID 21986246
IUPAC Name 3-bromo-5-fluorobenzaldehyde
SMILES C1=C(C=C(C=C1F)Br)C=O

Specifications

Melting Point 41°C to 43°C
Density 1.43 g/mL
Quantity 100 g
Beilstein 13904428
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 203.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Bromo-5-fluorobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.