missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-cyanopyridine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-5-cyanopyridine |
| CAS | 35590-37-5 |
| Molecular Formula | C6 H3 Br N2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H3BrN2/c7-6-1-5(2-8)3-9-4-6/h1,3-4H |
| SMILES | Brc1cncc(c1)C#N |
| IUPAC Name | 5-bromopyridine-3-carbonitrile |
| Molecular Weight (g/mol) | 183.01 |
| Formula Weight | 181.948 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction