missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chloro-2-hydroxybenzaldehyde, 97%

Product Code. 11435513
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435513 25 g 25g
11425513 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435513 Supplier Thermo Scientific Alfa Aesar Supplier No. A13449.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19652-32-5 | C7H4BrClO2 | 235.461 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19652-32-5
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00051690
InChI Key KNOYZLVIXXBBIB-UHFFFAOYSA-N
Synonym 3-bromo-5-chlorosalicylaldehyde, 3-bromo-5-chlorosalicylic aldehyde, benzaldehyde, 3-bromo-5-chloro-2-hydroxy, 3-bromo-5-chloro salicyclic aldehyde, 3-bromo-5-chloro-2-hydroxy-benzaldehyde, 3-bromo-5-chloro-o-hydroxybenzaldehyde, pubchem3591, acmc-1bs9s, akos bb-6948, knoyzlvixxbbib-uhfffaoysa
PubChem CID 519676
IUPAC Name 3-bromo-5-chloro-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Br)O)C=O)Cl

Specifications

Melting Point 84°C to 88°C
Quantity 25 g
Beilstein 2362159
Sensitivity Air Sensitive
Formula Weight 235.47
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-chloro-2-hydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.