missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylpyridine, 97%

Product Code. 10320144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10320144 5 g Glass bottle 5g
10433745 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10320144 Supplier Thermo Scientific Acros Supplier No. 392320050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3430-22-6 | C6H6BrN | 172.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3430-22-6
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.02
MDL Number MFCD00082592
InChI Key GSQZOLXWFQQJHJ-UHFFFAOYSA-N
Synonym 3-bromo-4-picoline, 4-methyl-3-bromopyridine, pyridine, 3-bromo-4-methyl, 3-bromo-4-methyl-pyridine, 3-brom-4-methylpyridin, zlchem 494, pubchem3994, 3-bromo4-methylpyridine, 3-bromo-4-methylpyridin
PubChem CID 817630
IUPAC Name 3-bromo-4-methylpyridine
SMILES CC1=C(C=NC=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5490g/mL
Boiling Point 76.0°C (10.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5600 to 1.5620
Quantity 5 g
Specific Gravity 1.549
Formula Weight 172.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.