missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-methylphenol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-4-methylphenol |
| CAS | 60710-39-6 |
| Molecular Formula | C7H7BrO |
| Quantity | 1 g |
| Synonym | 2-Bromo-4-hydroxytoluene, 3-Bromo-4-methylphenol, 2-Bromo-4-hydroxytoluene |
| InChI Formula | InChI=1S/C7H7BrO/c1-5-2-3-6(9)4-7(5)8/h2-4,9H,1H3 |
| SMILES | BrC1=C(C)C=CC(O)=C1 |
| IUPAC Name | 3-bromo-4-methylphenol |
| Molecular Weight (g/mol) | 187.03 |
| Formula Weight | 185.968 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction