missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylphenol, 98%

Product Code. p-7030430
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15443827

Brand: Thermo Scientific Alfa Aesar H28689.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 60710-39-6 | C7H7BrO | 187.036 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60710-39-6
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD08273793
InChI Key GMZKNRDHSHYMHG-UHFFFAOYSA-N
Synonym 3-bromo-p-cresol, 2-bromo-4-hydroxytoluene, phenol, 3-bromo-4-methyl, 3-bromo-4-methyl-phenol, 3-brom-p-kresol, acmc-209mlm, 3-bromo-4-methyl phenol, phenol,3-bromo-4-methyl, 3-bromanyl-4-methyl-phenol, ksc494c1p
PubChem CID 10307913
IUPAC Name 3-bromo-4-methylphenol
SMILES CC1=C(C=C(C=C1)O)Br

Specifications

Melting Point 54°C to 57°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 187.04
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-methylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.