missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylbenzoic acid, 98+%

Product Code. 11490403
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490403 25 g 25g
11480403 5 g 5g
11400413 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490403 Supplier Thermo Scientific Alfa Aesar Supplier No. A11496.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7697-26-9 | C8H7BrO2 | 215.05 g/mol

3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7697-26-9
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00002488
InChI Key ZFJOMUKPDWNRFI-UHFFFAOYSA-N
Synonym 3-bromo-4-methylbenzoicacid, 3-bromo-p-toluic acid, benzoic acid, 3-bromo-4-methyl, 3-bromo-4-methyl-benzoic acid, bromo-p-toluic acid, pubchem3806, bromo-para-toluic acid, 3-bromo4-methylbenzoic acid, acmc-1bg95, akos bbb/219
PubChem CID 82130
IUPAC Name 3-bromo-4-methylbenzoic acid
SMILES CC1=CC=C(C=C1Br)C(O)=O

Spécification

Melting Point 206°C to 211°C
Odor Odorless
Quantity 25 g
Beilstein 1936510
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-methylbenzoic acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.