missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxypyridine, 97%

Product Code. 15441429
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441429 5 g 5g
15451429 250 mg 250mg
15431429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441429 Supplier Thermo Scientific Alfa Aesar Supplier No. H64296.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82257-09-8 | C6H6BrNO | 188.024 g/mol

3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water (9.8 g/L).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82257-09-8
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD05664024
InChI Key DSFJKFHIMBVWAV-UHFFFAOYSA-N
PubChem CID 642827
IUPAC Name 3-bromo-4-methoxypyridine
SMILES COC1=C(C=NC=C1)Br

Specifications

Color Yellow
Quantity 5 g
Beilstein 1423610
Synonym 3-bromo-4-methoxy-pyridine, pyridine, 3-bromo-4-methoxy, pyridine,3-bromo-4-methoxy, pubchem6686, 4-methoxy-3-bromopyridine, acmc-209po0, abbypharma ap-17-5958
Solubility Information Slightly soluble in water (9.8g/L).
Formula Weight 188.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methoxypyridine

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.