missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybiphenyl, 99%, Thermo Scientific™

Product Code. 11490050
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11490050 250 mg 250mg
11400060 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11490050

Brand: Thermo Scientific Alfa Aesar H50581.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74447-73-7
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.13
MDL Number MFCD00079720
InChI Key QJGJVVIDRMVQMR-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxybiphenyl, 3-bromo-4-methoxy-1,1'-biphenyl, 2-bromo-1-methoxy-4-phenyl-menzene, acmc-20an8p, 2-bromo-4-phenylanisole, maybridge3_000975, 3-bromo-4-methoxy-biphenyl, 2-methoxy-5-phenyl-phenylbromide, 2-bromo-1-methoxy-4-phenyl-benzene
PubChem CID 262267
IUPAC Name 2-bromo-1-methoxy-4-phenylbenzene
SMILES COC1=CC=C(C=C1Br)C1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 263.14
Percent Purity 99%
Chemical Name or Material 3-Bromo-4-methoxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.