missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxy-5-methoxybenzaldehyde 97%, Maybridge™

Product Code. 10351453
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10351453

Brand: Thermo Scientific Maybridge BTB06293EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2973-76-4
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD00006940
InChI Key KLSHZDPXXKAHIJ-UHFFFAOYSA-N
Synonym 5-bromovanillin, benzaldehyde, 3-bromo-4-hydroxy-5-methoxy, vanillin, 5-bromo, 5-bromo-4-hydroxy-3-methoxybenzaldehyde, 3-bromo-4-hydroxy-5-methoxy-benzaldehyde, unii-508wvs56pw, 3-bromo-4-hydroxy-5-methyloxy benzaldehyde, 5-bromovaniline, 5-bromovanilline
PubChem CID 18099
IUPAC Name 3-bromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(Br)=C1O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula BrC6H2(OH)(OCH3)CHO
Quantity 10 g
Formula Weight 231.04
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxybenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.