missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-heptanone, 98%

Product Code. 11300046
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11300046

Brand: Thermo Scientific Alfa Aesar H32094.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42330-10-9 | C7H13BrO | 193.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42330-10-9
Molecular Formula C7H13BrO
Molecular Weight (g/mol) 193.08
MDL Number MFCD09757526
InChI Key SFKVBRLKXVRUQW-UHFFFAOYNA-N
Synonym 3-bromo-4-heptanone, 4-heptanone, 3-bromo
PubChem CID 11469593
IUPAC Name 3-bromoheptan-4-one
SMILES CCCC(=O)C(Br)CC

Specifications

Density 1.248
Boiling Point 39°C (1 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.458
Quantity 1 g
UN Number UN3265
Formula Weight 193.08
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-heptanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.