missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11409546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409546 25 g 25g
11499536 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409546 Supplier Thermo Scientific Alfa Aesar Supplier No. A19280.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-68-7
Molecular Formula C7H8BrClFN
Molecular Weight (g/mol) 240.50
MDL Number MFCD00143094
InChI Key RSFNYNQGERWCMT-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzylamine hydrochloride, 3-bromo-4-fluorophenyl methanamine hydrochloride, 3-bromo-4-fluorobenzylamine hcl, 1-3-bromo-4-fluorophenyl methanamine hydrochloride, 3-bromo-4-fluorophenyl methylamine, chloride, 1-3-bromo-4-fluorophenyl methylamine hydrochloride, benzenemethanamine, 3-bromo-4-fluoro-, hydrochloride, pubchem3226, 3-bromo-4-fluorobenzylaminehydrochloride, acmc-1bczm
PubChem CID 2724901
IUPAC Name (3-bromo-4-fluorophenyl)methanamine;hydrochloride
SMILES Cl.NCC1=CC(Br)=C(F)C=C1

Specifications

Melting Point 214°C to 218°C
Quantity 25 g
Formula Weight 240.5
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-fluorobenzylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.