missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11460571
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460571 25 g 25g
11440571 1 g 1g
11450571 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460571 Supplier Thermo Scientific Alfa Aesar Supplier No. B20078.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68322-84-9
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD00040938
InChI Key RZJOIMPUMMQKFR-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzotrifluoride, 2-bromo-1-fluoro-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl bromobenzene, 3-bromo-alpha,alpha,alpha,4-tetrafluorotoluene, benzene, 2-bromo-1-fluoro-4-trifluoromethyl, 1-bromo-2-fluoro-5-trifluoromethyl benzene, 3-bromo-alpha,alpha,alpha-4-tetrafluorotoluene, pubchem1639, 3-bromo-4-fluoro trifluoromethyl benzene
PubChem CID 144295
IUPAC Name 2-bromo-1-fluoro-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1Br)C(F)(F)F

Specifications

Density 1.71
Boiling Point 148°C to 149°C
Flash Point 71°C (159°F)
Refractive Index 1.461
Quantity 25 g
Beilstein 2093911
Formula Weight 243
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-fluorobenzotrifluoride
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.