missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10112814
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1007-16-5
C7H3BrFO2
218.00
MFCD00042463
ONELILMJNOWXSA-UHFFFAOYSA-M
3-bromo-4-fluoro-benzoic acid, 4-fluoro-3-bromo benzoic acid, 3-bromo-4-fluorobenzoicacid, rarechem al bo 0604, buttpark 20\01-58, benzoic acid, 3-bromo-4-fluoro, 3-bromo-4-fluorbenzoic acid, pubchem1320, 3-bromo-4-fiuorobenzoic acid, 5-bromo-4-fluorobenzoic acid
70509
[O-]C(=O)C1=CC=C(F)C(Br)=C1
This item is not returnable. View return policy
1007-16-5
C7H3BrFO2
218.00
MFCD00042463
ONELILMJNOWXSA-UHFFFAOYSA-M
3-bromo-4-fluoro-benzoic acid, 4-fluoro-3-bromo benzoic acid, 3-bromo-4-fluorobenzoicacid, rarechem al bo 0604, buttpark 20\01-58, benzoic acid, 3-bromo-4-fluoro, 3-bromo-4-fluorbenzoic acid, pubchem1320, 3-bromo-4-fiuorobenzoic acid, 5-bromo-4-fluorobenzoic acid
70509
[O-]C(=O)C1=CC=C(F)C(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 138.0°C to 140.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H3(F)COOH
Quantity 5 g
IUPAC Name 3-bromo-4-fluorobenzoate
Formula Weight 219
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-fluorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'