missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluoroaniline, 98%

Product Code. 11399899
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399899 25 g 25g
15413297 1 g 1g
11389899 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399899 Supplier Thermo Scientific Alfa Aesar Supplier No. B25610.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 656-64-4 | C6H5BrFN | 190.015 g/mol

3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 656-64-4
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD03788559
InChI Key KOWPUNQBGWIERF-UHFFFAOYSA-N
Synonym 3-bromo-4-fluoro aniline, benzenamine, 3-bromo-4-fluoro, 3-bromo-4-fluorobenzenamine, 3-bromo-4-fluorophenylamine, 3-bromo-4-fluoro-aniline, 3-bromo-4-fluoro-phenylamine, pubchem2924, acmc-209nrn, 3-bromo-4-fluoraniline, 3-bromo 4-fluoro-aniline
PubChem CID 2779286
IUPAC Name 3-bromo-4-fluoroaniline
SMILES C1=CC(=C(C=C1N)Br)F

Specifications

Melting Point 34°C to 36°C
Boiling Point 235°C
Quantity 25 g
UN Number UN2811
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-fluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.