missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chlorobenzotrifluoride, 98+%, Thermo Scientific™

Product Code. 10190402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10190402 5 g Glass bottle 5g
10376970 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10190402 Supplier Thermo Scientific Acros Supplier No. 137510050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 454-78-4
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00018093
InChI Key APSISOSWYXCEQX-UHFFFAOYSA-N
Synonym 2-bromo-1-chloro-4-trifluoromethyl benzene, 3-bromo-4-chlorobenzotrifluoride, benzene, 2-bromo-1-chloro-4-trifluoromethyl, 3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene, 4-chloro-3-bromobenzotrifluoride, 3-bromo-4-chloro-a,a,a-trifluorotoluene, pubchem4837, ksc915a8j, apsisoswyxceqx-uhfffaoysa, 3-bromo-4-chloro-benzotrifluoride
PubChem CID 67988
IUPAC Name 2-bromo-1-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -23°C to 22°C
Color Yellow
Density 1.7400g/mL
Boiling Point 188°C to 190°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula BrC6H3(Cl)CF3
Refractive Index 1.4980 to 1.5
Quantity 5 g
Specific Gravity 1.74
Formula Weight 259.45
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-chlorobenzotrifluoride, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-chlorobenzotrifluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.