missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chlorobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 10758678 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10758678 1 g 1g
10126107 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10758678 Supplier Thermo Scientific Acros Supplier No. 429870010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42860-10-6
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD00079706
InChI Key NLEPZGNUPNMRGF-UHFFFAOYSA-N
Synonym 3-bromo-4-chloro-benzoic acid, benzoic acid, 3-bromo-4-chloro, 4-chloro-3-bromobenzoic acid, pubchem3585, acmc-1cufe, 3-bromo-4-chlorobenzoicacid, ksc235m9l, timtec-bb sbb017622, zerenex e/9071912
PubChem CID 2735544
IUPAC Name 3-bromo-4-chlorobenzoic acid
SMILES OC(=O)C1=CC(Br)=C(Cl)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 220.0°C to 222.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 235.47
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-chlorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.