missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4,5-dimethoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 15274794
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6948-30-7
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
InChI Key ICVODPFGWCUVJC-UHFFFAOYSA-N
PubChem CID 81379
IUPAC Name 3-bromo-4,5-dimethoxybenzaldehyde
SMILES COC1=C(C(=CC(=C1)C=O)Br)OC

Specifications

Melting Point 61°C to 63°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 5-bromoveratraldehyde, 3-bromo-4,5-dimethoxy benzaldehyde, 3,4-dimethoxy-5-bromobenzaldehyde, benzaldehyde,3-bromo-4,5-dimethoxy, benzaldehyde, 3-bromo-4,5-dimethoxy, 5-bromoveratric aldehyde, acmc-209ml7, cambridge id 7187634, 5-bromveratrumaldehyde
Solubility Information Solubility in water: insoluble
Formula Weight 245.07
Percent Purity 97%
Chemical Name or Material 3-Bromo-4, 5-dimethoxybenzaldehyde
compliance-icons
Product Identifier
  • 3-Bromo-4,5-dimethoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'