missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-3-buten-1-ol, 97+%

Product Code. 10329143 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10329143 50 g 50g
10734563 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10329143 Supplier Thermo Scientific Acros Supplier No. 368210500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76334-36-6 | C4H7BrO | 151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76334-36-6
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151
MDL Number MFCD00154041
InChI Key RTKMFQOHBDVEBC-UHFFFAOYSA-N
Synonym 3-bromo-3-buten-1-ol, 3-buten-1-ol, 3-bromo, 3bb, zlchem 79, acmc-20an90, 3-bromanylbut-3-en-1-ol
PubChem CID 533975
IUPAC Name 3-bromobut-3-en-1-ol
SMILES C=C(CCO)Br

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Brown
Density 1.5220g/mL
Boiling Point 175.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula CH2=C(Br)CH2CH2OH
Refractive Index 1.4949
Quantity 50 g
Beilstein 01, IV, 2100
Specific Gravity 1.522
Formula Weight 151
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 3-Bromo-3-buten-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-3-buten-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.