missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylbenzoic acid, 97%

Product Code. 15461668
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461668 1 g 1g
15471668 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461668 Supplier Thermo Scientific Alfa Aesar Supplier No. H55436.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76006-33-2 | C8H7BrO2 | 215.05 g/mol

3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

Solubility
Soluble in chloroform, methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76006-33-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00270097
InChI Key BJGKVCKGUBYULR-UHFFFAOYSA-N
Synonym 3-bromo-o-toluic acid, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methyl-benzoic acid, benzoic acid, 3-bromo-2-methyl, pubchem4717, bromo-methylbenzoic acid, 2-bromo-6-carboxytoluene, 3-bromo-2-methybenzoic acid, 3-bromo-2-methylbenzoicacid, ksc377a4p
PubChem CID 2735590
IUPAC Name 3-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1Br)C(O)=O

Specifications

Melting Point 152°C to 156°C
Quantity 1 g
Solubility Information Soluble in chloroform,methanol
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.