missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylbenzoic acid, 97%

Product Code. p-7036946
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15461668

Brand: Thermo Scientific Alfa Aesar H55436.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76006-33-2 | C8H7BrO2 | 215.05 g/mol

3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

Solubility
Soluble in chloroform, methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76006-33-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00270097
InChI Key BJGKVCKGUBYULR-UHFFFAOYSA-N
Synonym 3-bromo-o-toluic acid, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methyl-benzoic acid, benzoic acid, 3-bromo-2-methyl, pubchem4717, bromo-methylbenzoic acid, 2-bromo-6-carboxytoluene, 3-bromo-2-methybenzoic acid, 3-bromo-2-methylbenzoicacid, ksc377a4p
PubChem CID 2735590
IUPAC Name 3-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1Br)C(O)=O

Specifications

Melting Point 152°C to 156°C
Quantity 1 g
Solubility Information Soluble in chloroform,methanol
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-methylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.