missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylaniline, 98%, Thermo Scientific™

Product Code. 11955221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11955221 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11955221 Supplier Thermo Scientific Acros Supplier No. 437810050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 55289-36-6
CAS Min % 97.5
CAS Max % 100.0
Color Brown to Colorless
Density 1.5100g/mL
Boiling Point 248°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8BrN
Quantity 5 mL
Specific Gravity 1.51
Synonym 2-amino-6-bromotoluene, 3-bromo-o-toluidine, benzenamine, 3-bromo-2-methyl, 3-bromo-2-methylbenzenamine, 2-bromo-6-aminotoluene, 3-bromo-2-methyl-phenylamine, 3-bromo-2-methyl-aniline, 3-bromo-2-methylphenylamine, 3-bromo-2-methyl-benzenamine, 3-brom-2-methylanilin
InChI Key IILVSKMKMOJHMA-UHFFFAOYSA-N
SMILES CC1=C(C=CC=C1Br)N
IUPAC Name 3-bromo-2-methylaniline
Molecular Weight (g/mol) 186.05
PubChem CID 123538
Formula Weight 186.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-2-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.