missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylaniline, 98+%

Product Code. 11473854
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473854 100 g 100g
11453854 5 g 5g
11463854 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473854 Supplier Thermo Scientific Alfa Aesar Supplier No. A11884.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 55289-36-6 | C7H8BrN | 186.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55289-36-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD00051579
InChI Key IILVSKMKMOJHMA-UHFFFAOYSA-N
Synonym 2-amino-6-bromotoluene, 3-bromo-o-toluidine, benzenamine, 3-bromo-2-methyl, 3-bromo-2-methylbenzenamine, 2-bromo-6-aminotoluene, 3-bromo-2-methyl-phenylamine, 3-bromo-2-methyl-aniline, 3-bromo-2-methylphenylamine, 3-bromo-2-methyl-benzenamine, 3-brom-2-methylanilin
PubChem CID 123538
IUPAC Name 3-bromo-2-methylaniline
SMILES CC1=C(C=CC=C1Br)N

Specifications

Density 1.51
Boiling Point 253°C to 255°C
Flash Point >110°C (230°F)
Refractive Index 1.616
Quantity 100 g
UN Number UN2810
Beilstein 2354213
Sensitivity Light sensitive
Formula Weight 186.06
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-2-methylaniline
Show More Show Less

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.